Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bli-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067371.3 | NP_499772.1 | 2 - 3 | 263 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |