Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sid-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067422.7 | NP_499823.2 | 3 - 4 | 716 | 118 | |
GenBank mRNA | AY466439.1 | - | 3 - 4 | 738 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |