Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frm-5.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268287.2 | NP_001255216.1 | 1 - 2 | 90 | 76 | |
NM_001268288.4 | NP_001255217.2 | 1 - 2 | 90 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |