Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Y38C1AA.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379994.1 | NP_001367798.1 | 8 - 9 | 2892 | 113 |
Gene Symbol: cutl-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028278.5 | NP_001023449.1 | 8 - 9 | 2903 | 113 | |
NM_001028279.6 | NP_001023450.1 | 6 - 7 | 1334 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |