Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067520.9 | NP_499921.2 | 2 - 3 | 274 | 66 | |
GenBank mRNA | AY008127.1 | - | 2 - 3 | 235 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |