Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067520.9 | NP_499921.2 | 6 - 7 | 1001 | 146 | |
GenBank mRNA | AY008127.1 | - | 6 - 7 | 962 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |