Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067525.6 | NP_499926.1 | 5 - 6 | 2410 | 73 | |
GenBank mRNA | AF098286.1 | - | 5 - 6 | 2402 | 73 | |
AF126286.1 | - | 5 - 6 | 2401 | 73 | ||
AJ131181.1 | - | 5 - 6 | 2401 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |