Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307363.3 | NP_001294292.1 | 17 - 18 | 1914 | 82 | |
NM_001307364.3 | NP_001294293.1 | 16 - 17 | 1920 | 82 | ||
NM_001307365.3 | NP_001294294.1 | 15 - 16 | 1797 | 82 | ||
NM_068737.2 | NP_501138.1 | 17 - 18 | 1917 | 82 | ||
GenBank mRNA | U76402.1 | - | 17 - 18 | 1917 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |