Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027970.4 | NP_001023141.1 | 2 - 3 | 131 | 76 | |
NM_001027971.6 | NP_001023142.1 | 2 - 3 | 482 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |