Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rig-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068938.8 | NP_501339.2 | 5 - 6 | 1178 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |