Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fem-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268436.3 | NP_001255365.1 | 6 - 7 | 937 | 83 | |
GenBank mRNA | X64962.1 | - | 330 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |