Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136399.3 | NP_001129871.1 | 4 - 5 | 865 | 66 | |
NM_001276834.3 | NP_001263763.1 | 4 - 5 | 1027 | 66 | ||
NM_001380407.1 | NP_001368165.1 | 5 - 6 | 990 | 66 | ||
NM_069243.5 | NP_501644.1 | 4 - 5 | 1021 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |