Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: amph-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069310.5 | NP_501711.1 | 12 - 13 | 1261 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |