Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gon-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268519.3 | NP_001255448.1 | 21 - 22 | 5109 | 77 | |
NM_001372913.1 | NP_001359558.1 | 23 - 24 | 5568 | 77 | ||
NR_056474.1 | 23 - 24 | 5564 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |