Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gst-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069447.7 | NP_501848.1 | 2 - 3 | 333 | 80 | |
GenBank mRNA | AF010239.1 | - | 2 - 3 | 228 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |