Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306925.2 | NP_001293854.1 | 13 - 14 | 2229 | 95 | |
NM_001306926.3 | NP_001293855.1 | 10 - 11 | 1992 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |