Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268546.3 | NP_001255475.1 | 3 - 4 | 304 | 142 | |
NM_001268547.2 | NP_001255476.1 | 1 - 2 | 153 | 142 | ||
GenBank mRNA | AF190910.1 | - | 2 - 3 | 193 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |