Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028127.6 | NP_001023298.1 | 7 - 8 | 906 | 76 | |
NM_001268551.3 | NP_001255480.1 | 8 - 9 | 925 | 76 | ||
NM_001372918.2 | NP_001359633.1 | 8 | 986 | 76 | ||
NM_001380439.2 | NP_001366993.1 | 8 - 9 | 1046 | 76 | ||
NM_001380440.2 | NP_001366994.1 | 8 - 9 | 1000 | 76 | ||
NM_001380441.2 | NP_001366996.1 | 5 - 6 | 749 | 76 | ||
NM_001380442.1 | NP_001366992.1 | 6 - 7 | 830 | 76 | ||
NM_001380443.1 | NP_001366997.1 | 5 - 6 | 595 | 76 | ||
NM_001383186.1 | 7 - 8 | 906 | 76 | |||
NM_001383187.1 | 7 - 8 | 907 | 76 | |||
GenBank mRNA | AF180735.1 | - | 6 - 7 | 808 | 76 | |
AF233262.1 | - | 5 - 6 | 595 | 76 | ||
AF233263.1 | - | 5 - 6 | 595 | 76 | ||
AF233264.1 | - | 5 - 6 | 595 | 76 | ||
AF233265.1 | - | 5 - 6 | 595 | 76 | ||
AF233266.1 | - | 5 - 6 | 595 | 76 | ||
AF233267.1 | - | 5 - 6 | 595 | 76 | ||
AF255956.1 | - | 5 - 6 | 595 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |