Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268558.2 | NP_001255487.1 | 2 - 3 | 347 | 93 | |
NM_001268559.3 | NP_001255488.1 | 2 - 3 | 277 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |