Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268558.2 | NP_001255487.1 | 5 - 6 | 1057 | 75 | |
NM_001268559.3 | NP_001255488.1 | 5 - 6 | 987 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |