Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028110.4 | NP_001023281.1 | 10 - 11 | 8974 | 115 | |
NM_001028111.5 | NP_001023282.1 | 10 - 11 | 8962 | 115 | ||
NM_001268571.3 | NP_001255500.1 | 11 - 12 | 9001 | 115 | ||
NM_001268572.3 | NP_001255501.1 | 10 - 11 | 8941 | 115 | ||
NM_001307078.4 | NP_001294007.1 | 10 - 11 | 8945 | 115 | ||
NM_001307079.3 | NP_001294008.1 | 11 - 12 | 8995 | 115 | ||
GenBank mRNA | AB001074.1 | - | 10 - 11 | 8968 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |