Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268592.3 | NP_001255521.1 | 12 - 13 | 3458 | 99 | |
NM_001268593.3 | NP_001255522.1 | 6 - 7 | 2571 | 99 | ||
NM_001276852.4 | NP_001263781.1 | 10 - 11 | 3256 | 99 | ||
NM_001276853.3 | NP_001263782.1 | 11 - 12 | 3374 | 99 | ||
NM_001383200.1 | 12 - 13 | 3503 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |