Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069666.6 | NP_502067.2 | 14 - 15 | 1897 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |