Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asm-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047531.2 | NP_001040996.1 | 4 - 5 | 446 | 70 | |
NM_001047532.6 | NP_001040997.1 | 3 - 4 | 244 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |