Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067770.5 | NP_500171.2 | 5 - 6 | 541 | 110 | |
GenBank mRNA | AY652946.1 | - | 5 - 6 | 534 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |