Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028336.4 | NP_001023507.1 | 9 - 10 | 1379 | 64 | |
NM_001383059.1 | 9 - 10 | 1363 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |