Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067908.5 | NP_500309.3 | 3 - 4 | 640 | 68 | |
GenBank mRNA | AB201390.1 | - | 3 - 4 | 640 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |