Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: map-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_067995.4 | NP_500396.2 | 5 - 6 | 885 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |