Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068031.6 | NP_500432.3 | 3 - 4 | 540 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |