Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpf-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380210.1 | NP_001368236.1 | 5 - 6 | 1645 | 82 | |
NM_068246.7 | NP_500647.2 | 6 - 7 | 2128 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |