Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srm-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068300.2 | NP_500701.2 | 5 - 6 | 901 | 75 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |