Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fem-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068424.4 | NP_500825.1 | 4 - 5 | 361 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |