Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: best-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383107.1 | 3 - 4 | 285 | 123 |
Gene Symbol: try-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306808.3 | NP_001293737.1 | 2 - 3 | 233 | 123 | |
NM_001380280.2 | NP_001367393.1 | 3 - 4 | 458 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |