Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276863.2 | NP_001263792.1 | 1 - 2 | 123 | 106 | |
NM_069769.5 | NP_502170.4 | 10 - 11 | 1137 | 106 | ||
NR_101961.1 | 10 - 11 | 1085 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |