Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mep-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069772.8 | NP_502173.2 | 4 - 5 | 1903 | 76 | |
GenBank mRNA | AF416567.1 | - | 4 - 5 | 1925 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |