Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lips-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083237.3 | NP_001076706.1 | 4 - 5 | 525 | 115 | |
NM_001268659.2 | NP_001255588.1 | 4 - 5 | 631 | 115 | ||
NM_001268660.3 | NP_001255589.1 | 4 - 5 | 528 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |