Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpa-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307154.4 | NP_001294083.1 | 5 - 6 | 1671 | 79 | |
NM_069848.7 | NP_502249.3 | 5 - 6 | 1668 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |