Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129363.4 | NP_001122835.2 | 11 - 12 | 1906 | 69 | |
NM_001268672.3 | NP_001255601.1 | 17 - 18 | 2569 | 69 | ||
NM_001268674.4 | NP_001255603.2 | 8 - 9 | 1144 | 69 | ||
NM_001268675.3 | NP_001255604.1 | 7 - 8 | 1093 | 69 | ||
NM_001307046.3 | NP_001293975.1 | 19 - 20 | 3187 | 69 | ||
NM_069872.6 | NP_502273.2 | 17 - 18 | 2432 | 69 | ||
NM_069873.6 | NP_502274.2 | 20 - 21 | 3262 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |