Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sre-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306971.3 | NP_001293900.1 | 1 - 2 | 108 | 135 | |
NM_001307201.4 | NP_001294130.1 | 6 - 7 | 768 | 135 | ||
NR_131614.1 | 5 - 6 | 610 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |