Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070011.5 | NP_502412.1 | 2 - 3 | 898 | 86 | |
NR_131627.1 | 3 - 4 | 793 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |