Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383222.1 | 17 - 18 | 2382 | 88 | ||
NM_070018.6 | NP_502419.2 | 12 - 13 | 1650 | 88 | ||
GenBank mRNA | AB055112.1 | - | 14 - 15 | 2047 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |