Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pin-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268743.2 | NP_001255672.1 | 3 - 4 | 371 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |