Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028095.6 | NP_001023266.1 | 5 - 6 | 1495 | 83 | |
GenBank mRNA | AF283322.1 | - | 5 - 6 | 1515 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |