Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsd-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047487.3 | NP_001040952.2 | 16 - 17 | 2844 | 119 | |
NM_001047488.4 | NP_001040953.1 | 13 - 14 | 2274 | 119 | ||
NM_001268820.3 | NP_001255749.1 | 9 - 10 | 1500 | 119 | ||
NM_001268821.3 | NP_001255750.1 | 1 - 2 | 138 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |