Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rsd-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047487.3 | NP_001040952.2 | 9 - 10 | 1786 | 129 | |
NM_001047488.4 | NP_001040953.1 | 6 - 7 | 1216 | 129 | ||
NM_001268820.3 | NP_001255749.1 | 2 - 3 | 442 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |