Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383253.1 | 10 - 11 | 1567 | 116 | ||
NR_101983.1 | 10 - 11 | 1501 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |