Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jac-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047542.3 | NP_001041007.1 | 2 - 3 | 224 | 63 | |
NM_001307194.3 | NP_001294123.1 | 2 - 3 | 224 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |