Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047601.5 | NP_001041066.1 | 6 - 7 | 1225 | 103 | |
NM_001380299.1 | NP_001367362.1 | 6 - 7 | 1156 | 103 | ||
GenBank mRNA | AF077868.1 | - | 5 - 6 | 1139 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |