Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047601.5 | NP_001041066.1 | 7 - 8 | 1401 | 87 | |
NM_001380299.1 | NP_001367362.1 | 7 - 8 | 1332 | 87 | ||
GenBank mRNA | AF077868.1 | - | 6 - 7 | 1315 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |