Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cutl-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_070984.5 | NP_503385.3 | 2 - 3 | 394 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |